Learn More
Enzo Life Sciences CaMKII monoclonal antibody (6G9) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCA002D
Encompass_Preferred
Clone: 6G9. Host: Mouse. Isotype: IgG1. Immunogen: Rat CaMKII (partially purified). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Mouse, Rat. Applications: IHC, WB in vitro Assay. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7229