Learn More
Enzo Life Sciences CaMKII polyclonal antibody (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPCA002F
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the N-terminus of rat CaMKII δ. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Hamster, Guinea pig, Chicken, Bovine. Applications: IHC (PS), WB. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7262