Learn More
Enzo Life Sciences CaMKII polyclonal antibody (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKAPCA002D
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the N-terminus of rat CaMKII δ. UniProt ID: P15791. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Rabbit, Hamster, Guinea pig, Chicken, Bovine. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (Colorimetric, 1:500) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7261