Learn More
Enzo Life Sciences Ca2+/calmodulin-dependent protein kinase II (rat) (alpha subunit) (pThr286) monoclonal antibody (22B1) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX802009R100
Encompass_Preferred
Clone: 22B1. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 281-294 (M281HRQET(PSO3)VDCLKKFN294) of the α-subunit of mouse and rat CaM kinase II (Ca2+/Calmodulin-dependent Protein Kinase II) thiophosphorylated at (Thr286). Species reactivity: Human, Mouse, Rat. Specificity: Recognizes the phosphorylated α-subunit of CaM kinase II. This antibody does not detect the non-phosphorylated α-subunit of rat CaM kinase II nor the ~60kDa β-subunit in either phosphorylation state. Applications: ELISA, Flow Cytometry, ICC, IF, IP, WB. Formulation: Liquid. In PBS containing 0.05% sodium azide.
Catalog No. 50-200-9721