Learn More
Aviva Systems Biology Corp CAMKK1 Antibody - C-terminal region (ARP60405_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60405_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAMKK1. It was validated on Western Blot. . Alternate names: CAMKKA, DKFZp761M0423, MGC34095ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-036