Learn More
Carnabio Usa Inc CAMKK2/5UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 051085
Encompass
CAMKK2, Full-length, Wild type, Amino Acid 1-588, NP_006540.3(isoform-1), Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0359