Learn More
Aviva Systems Biology Corp CAMKK2 Antibody - N-terminal region (ARP53184_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53184_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAMKK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CAMKK2. Alternate names: CAMKK, CAMKKB, KIAA0787, MGC15254ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60960