Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CAMKK2 (CaMKK Beta), GST Tag Recombinant Protein

Catalog No. PR7777A
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR7777A 100 μg
PV4322 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR7777A Supplier Invitrogen™ Supplier No. PR7777A
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length

CAMKK2 (CaMKK beta) is a serine/threonine kinase involved in neurogenesis, cell differentiation, and signal transduction. It also activates CAMKI and CAMKIV, and plays a role in the development of long term memory.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96RR4
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.02% Triton X-100, 50% glycerol, 2mM DTT, 150mM NaCl, 0.5mM EDTA and no preservative; pH 7.5
Gene ID (Entrez) 10645
Molecular Weight (g/mol) 87.1 kDa
Name Human CAMKK2 (CaMKK Beta), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 6330570N16Rik; AW061083; Ca+/Calmodulin-dependent protein kinase kinase beta (CaM-kinase kinase beta); calcium/calmodulin dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase 2 beta; calcium/calmodulin-dependent protein kinase beta; calcium/calmodulin-dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase kinase 2, beta; calcium/calmodulin-dependent protein kinase kinase beta; caM-kinase kinase 2; CaM-kinase kinase beta; CAMKK; caMKK 2; caM-KK 2; caMKK beta; CaM-KK beta; CAMKK beta protein; CAMKK2; CAMKKB; KIAA0787; MGC15254; mKIAA0787
Common Name CAMKK2
Gene Symbol CAMKK2
Kinase Family CAMKK Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage Storage Buffer: 50 mM Tris-HCl pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, and 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Kinase Group Ser⁄Thr Kinases: Other
Shipping Condition Dry Ice
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.