Learn More
Aviva Systems Biology Corp CAMLG Antibody - N-terminal region (ARP48191_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48191_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAMLG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CAMLG. Alternate names: CAML, MGC163197ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-59750