Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BoxLock CamLock with Lab Box - Includes 2 Year Software License

Catalog No. 22458000
Change view
Click to view available options
Color:
Lock: Yellow and Black; Box: White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-458-000 Lock: Yellow and Black; Box: White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-458-000 Supplier BoxLock Supplier No. FGV3BHCLKF100TF2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

With a built-in high-quality scanner, the BoxLock CamLock is ideal for automating specimen drop and retrieval. This combo includes a BoxLock CamLock pre-installed on a Small Specimen Drop Box. The bundle also includes 2 years of connectivity and access to BoxLock Control software.

  • Cam Lock on McShane KF100 11.5 (H) X 11.5 (W) X 9 (D) lab box with Zebra SE4107 scanner, 2.4 Ghz Wi-Fi and LTE-M/NB-IoT Connectivity, target IP-67 Rated, 1 Year Replaceable and Rechargeable Battery, Bluetooth IoT Gateway, Intrusion Detection.
  • Includes 2 Year of Software and Connectivity.
  • Each specimen box is made from 20 gauge galvanneal steel. Boxes come insulated for specimen integrity with 1/2 in. EPS foam.
  • New customers must also purchase Account Activation or the BoxLock Gold Service Plan before activating their devices.

Specifications

Color Lock: Yellow and Black; Box: White
Key Type Electronic/Scanner
Locking Mechanism Cam
Material Plastic and Steel
Product Type Electronic Cam Lock
For Use With (Application) Lab Specimen Drop/Retrieval
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.