Learn More
Medchemexpress LLC HY-14916 5mg Medchemexpress, Camobucol CAS:216167-92-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY149165MG
Encompass_Preferred
Medchemexpress, HY-14916 5mg Camobucol CAS:216167-92-9 Camobucol is an orally active, phenolic antioxidant and anti-inflammatory compound with antirheumatic properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9924