Learn More
Medchemexpress LLC Camonsertib | 2417489-10-0 | MFCD34578462 | 100.0% | C21H26N6O3 | 10MG
SDP
Catalog No. 5000261723
Encompass_Preferred
Camonsertib is an orally active, selective ATR kinase inhibitor developed for oncology research and clinical investigation. It exhibits low-nanomolar biochemical potency against ATR, reported selectivity versus related kinases, and has shown antitumor activity in preclinical and early clinical studies. The compound is supplied as a high-purity solid for research use only.
  • Potent ATR inhibition with low-nanomolar biochemical potency.
  • High selectivity versus related kinases, limiting off-target effects.
  • Demonstrated antitumor activity in preclinical and early clinical evaluations.
  • Provided as a high-purity solid suitable for laboratory research.
  • Available in small research-scale quantities for experimental studies.
Catalog No. 50-002-61723 Supplier Medchemexpress LLC Supplier No. HY13960910MG
Camonsertib is an orally active, selective ATR kinase inhibitor developed for oncology research and clinical investigation. It exhibits low-nanomolar biochemical potency against ATR, reported selectivity versus related kinases, and has shown antitumor activity in preclinical and early clinical studies. The compound is supplied as a high-purity solid for research use only.
  • Potent ATR inhibition with low-nanomolar biochemical potency.
  • High selectivity versus related kinases, limiting off-target effects.
  • Demonstrated antitumor activity in preclinical and early clinical evaluations.
  • Provided as a high-purity solid suitable for laboratory research.
  • Available in small research-scale quantities for experimental studies.