Learn More
Selleck Chemical LLC Camonsertib-E1108-10MM/1ML
SDP
Catalog No. 503136761
Encompass_Preferred
Camonsertib is a highly potent and selective inhibitor of ATR kinase with an IC50 of 1 nM
Catalog No. 50-313-6761 Supplier Selleck Chemical LLC Supplier No. E110810MM/1ML
Add to Cart
Add to Cart
Camonsertib is a highly potent and selective inhibitor of ATR kinase with an IC50 of 1 nM