Learn More
Selleck Chemical LLC Camonsertib-E1108-5MG
SDP
Catalog No. 502389847
Encompass_Preferred
Camonsertib is a highly potent and selective inhibitor of ATR kinase with an IC50 of 1 nM
Catalog No. 50-238-9847 Supplier Selleck Chemical LLC Supplier No. E11085MG
Camonsertib is a highly potent and selective inhibitor of ATR kinase with an IC50 of 1 nM