Learn More
TARGETMOL CHEMICALS INC Camostat mesylate 100MG
SDP
Catalog No. 502877760
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Camostat mesylate (FOY-S980) is a trypsin-like protease inhibitor and inhibits airway epithelial sodium channel (ENaC) function. Purity 99.69%
Catalog No. 50-287-7760 Supplier TARGETMOL CHEMICALS INC Supplier No. T2391100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Camostat mesylate (FOY-S980) is a trypsin-like protease inhibitor and inhibits airway epithelial sodium channel (ENaC) function. Purity 99.69%