Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Camostat mesylate | 59721-29-8 | MFCD00941410 | 100.0% | C21H26N4O8S | 10MG
SDP

Catalog No. 5000260529
Encompass_Preferred

Camostat mesylate is an orally active synthetic serine protease inhibitor used in biochemical and antiviral research. It inhibits trypsin-like proteases including TMPRSS2 and is applied in studies of protease function, viral entry inhibition, and inflammatory pathways. Supplied as a high-purity standard for laboratory assays and pharmacological research.

  • Targets TMPRSS2 and trypsin-like serine proteases.
  • High purity suitable for analytical and biological assays (reported 99.96%).
  • Molecular formula C21H26N4O8S and molecular weight 494.52.
  • Supplied as a solid reference material for in vitro and biochemical studies.
  • Relevant for antiviral research, protease inhibition assays, and inflammation studies.

Catalog No. 50-002-60529 Supplier Medchemexpress LLC Supplier No. HY13512R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Camostat mesylate is an orally active synthetic serine protease inhibitor used in biochemical and antiviral research. It inhibits trypsin-like proteases including TMPRSS2 and is applied in studies of protease function, viral entry inhibition, and inflammatory pathways. Supplied as a high-purity standard for laboratory assays and pharmacological research.

  • Targets TMPRSS2 and trypsin-like serine proteases.
  • High purity suitable for analytical and biological assays (reported 99.96%).
  • Molecular formula C21H26N4O8S and molecular weight 494.52.
  • Supplied as a solid reference material for in vitro and biochemical studies.
  • Relevant for antiviral research, protease inhibition assays, and inflammation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.