Learn More
Enzo Life Sciences cAMP Complete ELISA Kit, Quantitative Cyclic AMP Measurement, 96 Wells
SDP
Supplier:  Enzo Life Sciences ADI900163A
Encompass_Preferred
The Enzo Life Sciences cAMP Complete ELISA Kit is a competitive immunoassay designed for quantitative measurement of cyclic AMP in diverse biological samples. It features an optional acetylation protocol that enhances sensitivity by approximately 10-fold. This kit allows for direct measurement with simplified sample preparation without extraction steps, making it ideal for samples with low cAMP levels. Its colorimetric detection method provides accurate results at 405 nm.
Catalog No. NC2068022