Learn More
New England Biolabs, Inc. cAMP-dependent Protein Kinase (PKA), catalytic subunit – 500000 units
SDP
Catalog No. 504440
Encompass
The catalytic subunit of cAMP-dependent Protein Kinase (PKA) is a serine/threonine protein kinase. The recognition motif for phosphorylation by PKA is RRXS/TY, where Y tends to be a hydrophobic residue.
Catalog No. 50-444-0 Supplier New England Biolabs, Inc. Supplier No. P6000L
The catalytic subunit of cAMP-dependent Protein Kinase (PKA) is a serine/threonine protein kinase. The recognition motif for phosphorylation by PKA is RRXS/TY, where Y tends to be a hydrophobic residue.