Learn More
CPC Scientific H-Thr-Thr-Tyr-Ala-Asp-Phe-Ile-Ala-Ser-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110158
Encompass
ONE-LETTER SEQUENCE: TTYADFIASGRTGRRNAIHD-NH2MOLECULAR FORMULA: C94H149N33O30MOLECULAR WEIGHT:2221.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: G. Swarup et al., JBC, 258, 10341 (1983)
Catalog No. 50-211-0158 Supplier CPC Scientific Supplier No. PKCP005A
ONE-LETTER SEQUENCE: TTYADFIASGRTGRRNAIHD-NH2MOLECULAR FORMULA: C94H149N33O30MOLECULAR WEIGHT:2221.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: G. Swarup et al., JBC, 258, 10341 (1983)