Learn More
CPC Scientific H-Thr-Thr-Tyr-Ala-Asp-Phe-Ile-Ala-Ser-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110157
Encompass
cAMP-dependent protein kinase inhibitor.ONE-LETTER SEQUENCE: TTYADFIASGRTGRRNAI-NH2MOLECULAR FORMULA: C84H137N29O26MOLECULAR WEIGHT:1969.19STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [100853-58-5], SYNONYMS: Wiptide, PKI-α (5-22) amide (human, mouse, rabbit, rat), cAMP-Dependent Protein Kinase Inhibitor- (5-22) amide (human, mouse, rabbit, rat)RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: H.C. Cheng et al., JBC, 261, 984 (1986)
Catalog No. 50-211-0157 Supplier CPC Scientific Supplier No. PKCP003A
cAMP-dependent protein kinase inhibitor.ONE-LETTER SEQUENCE: TTYADFIASGRTGRRNAI-NH2MOLECULAR FORMULA: C84H137N29O26MOLECULAR WEIGHT:1969.19STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [100853-58-5], SYNONYMS: Wiptide, PKI-α (5-22) amide (human, mouse, rabbit, rat), cAMP-Dependent Protein Kinase Inhibitor- (5-22) amide (human, mouse, rabbit, rat)RESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: H.C. Cheng et al., JBC, 261, 984 (1986)