Learn More
CPC Scientific H-Gly-Arg-Thr-Gly-Arg-Arg-Asn-Ala-Ile-His-Asp-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110159
Encompass
ONE-LETTER SEQUENCE: GRTGRRNAIHD-NH2MOLECULAR FORMULA: C49H86N24O15MOLECULAR WEIGHT:1251.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: H. C. Cheng et al., JBC, 261, 989 (1986)
Catalog No. 50-211-0159 Supplier CPC Scientific Supplier No. PKCP006A
ONE-LETTER SEQUENCE: GRTGRRNAIHD-NH2MOLECULAR FORMULA: C49H86N24O15MOLECULAR WEIGHT:1251.4STORAGE CONDITIONS: -20 5CRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES: H. C. Cheng et al., JBC, 261, 989 (1986)