Learn More
Raybiotech Inc cAMP EIA Kit.
Supplier:  Raybiotech Inc EIACAMP1
Encompass_Preferred
Optimized for serum and plasma samples. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8804