Learn More
Cell Biolabs Inc cAMP (Cyclic AMP) ELISA Kit (Chemiluminescent), 96 assays
Supplier:  Cell Biolabs Inc STA501
Encompass_Preferred
Cyclic AMP (cAMP) is a ubiquitous second messenger involved in various cellular activities in many cell and tissue types. Cell Biolabs’ cAMP ELISA Kit is a competitive enzyme immunoassay designed to measure cyclic AMP in cell culture supernatants, plasma, serum, saliva, urine, and cell lysates with chemiluminescent detection. The kit selectively measures cAMP levels without any significant cross reactivities to other nucleotides or cyclic nucleotides. Samples containing low cAMP levels may be acetylated (reagents provided) for increased sensitivity. Under non-acetylated conditions, the kit has a detection range of 1 to 1000 pmol/mL cAMP; however, under acetylated conditions, the sensitivity is enhanced (approx 100X) to a detection range of 10-2500 fmol/mL. Each kit contains sufficient reagents for 96 assays including standards and unknown samples.
Catalog No. NC0005227