Learn More
Enzo Life Sciences cAMP polyclonal antibody (100µl)
SDP
Supplier:  Enzo Life Sciences ADI915053100
Encompass_Preferred
Host: Rabbit. Immunogen: cAMP conjugated to a protein carrier molecule. Source: Whole rabbit serum. Species reactivity: Species independent. Applications: ELISA. Formulation: Neat serum.
Catalog No. 50-200-7106