Learn More
ABclonal Technology CAMP Rabbit mAb
SDP
Catalog No. 5000011525
Encompass_Preferred
This gene encodes a member of an antimicrobial peptide family characterized by a highly conserved N-terminal signal peptide containing a cathelin domain and a structurally variable cationic antimicrobial peptide which is produced by extracellular proteolysis from the C-terminus The protein plays an important role in innate immunity defense against viruses In addition to its antibacterial antifungal and antiviral activities the encoded protein functions in cell chemotaxis immune mediator induction and inflammatory response regulation
Catalog No. 50-000-11525 Supplier ABclonal Technology Supplier No. A2727120UL
This gene encodes a member of an antimicrobial peptide family characterized by a highly conserved N-terminal signal peptide containing a cathelin domain and a structurally variable cationic antimicrobial peptide which is produced by extracellular proteolysis from the C-terminus The protein plays an important role in innate immunity defense against viruses In addition to its antibacterial antifungal and antiviral activities the encoded protein functions in cell chemotaxis immune mediator induction and inflammatory response regulation