Learn More
MSC Campbell Anchor Shackle, Screw Pin, 1/2 in, Galvanized, 2 Ton WLL
Catalog No. 5000532127
Encompass
This screw pin anchor shackle is constructed from drop-forged carbon steel and hot-dip galvanized for enhanced durability and corrosion resistance. Designed for rigging applications, it features a 1/2 inch trade size and a working load limit of 2 short tons. The shackle bodies are statistically tested for impact resistance, tensile strength, hardness, ductility, and fatigue, ensuring reliable performance in demanding conditions.
  • Working load limit, size, and date code permanently marked.
  • Meets or exceeds all manufacturing requirements of ASME B30.26 "Rigging Hardware".
  • Meets design requirements of Federal Specification RR-C-271, Type IVA, Grade A, Class 2.
  • Galvanized to meet ASTM A-153 specifications C-419-G.
  • Pins and bolts are heat-treated and made of alloy steel.
  • Features a 6 to 1 design factor.
Catalog No. 50-005-32127 Supplier MSC Supplier No. 65088338
This screw pin anchor shackle is constructed from drop-forged carbon steel and hot-dip galvanized for enhanced durability and corrosion resistance. Designed for rigging applications, it features a 1/2 inch trade size and a working load limit of 2 short tons. The shackle bodies are statistically tested for impact resistance, tensile strength, hardness, ductility, and fatigue, ensuring reliable performance in demanding conditions.
  • Working load limit, size, and date code permanently marked.
  • Meets or exceeds all manufacturing requirements of ASME B30.26 "Rigging Hardware".
  • Meets design requirements of Federal Specification RR-C-271, Type IVA, Grade A, Class 2.
  • Galvanized to meet ASTM A-153 specifications C-419-G.
  • Pins and bolts are heat-treated and made of alloy steel.
  • Features a 6 to 1 design factor.