Learn More
MSC Campbell Cable, 250 ft, 3/16 in, 7 x 19 Strand, Galvanized, Uncoated
Catalog No. 5000530446
Encompass
This high-strength wire cable is suitable for a variety of demanding applications. It features a galvanized finish for enhanced durability and corrosion resistance, and its flexible construction makes it versatile for different uses.
  • Length: 250 feet
  • Diameter: 3/16 inch
  • Breaking strength of 840 pounds
  • 7 x 19 strand type construction
  • Uncoated for specific applications
  • Galvanized finish for durability
  • Flexible design for versatile use
  • Designed with a 5 to 1 design factor
Catalog No. 50-005-30446 Supplier MSC Supplier No. 95095600
This high-strength wire cable is suitable for a variety of demanding applications. It features a galvanized finish for enhanced durability and corrosion resistance, and its flexible construction makes it versatile for different uses.
  • Length: 250 feet
  • Diameter: 3/16 inch
  • Breaking strength of 840 pounds
  • 7 x 19 strand type construction
  • Uncoated for specific applications
  • Galvanized finish for durability
  • Flexible design for versatile use
  • Designed with a 5 to 1 design factor