Learn More
MSC Campbell Double Wood Shell Block, 4 in, 1/2 in Rope, 1400 lb WLL
Catalog No. 5000530635
Encompass
This double wood shell block is designed for various lifting and rigging applications. It features a 4 in block size, accommodates a 1/2 in rope, and has a working load limit of 1400 lb. Constructed with hard wood shells and galvanized metal parts, it ensures durability and reliability. The block is equipped with a swivel hook and bronze self-lubricating bushings for smooth operation.
  • Hard wood shells
  • Drop forged fittings
  • Bronze self-lubricating bushings
  • Galvanized metal parts
  • Accommodates 1/2 in rope diameter
  • 2-1/4 in outside diameter sheave
  • 4 in block size
  • Double number of sheaves
  • Swivel hook with latch end fitting
  • 1400 lb working load limit
Catalog No. 50-005-30635 Supplier MSC Supplier No. 67329441
This double wood shell block is designed for various lifting and rigging applications. It features a 4 in block size, accommodates a 1/2 in rope, and has a working load limit of 1400 lb. Constructed with hard wood shells and galvanized metal parts, it ensures durability and reliability. The block is equipped with a swivel hook and bronze self-lubricating bushings for smooth operation.
  • Hard wood shells
  • Drop forged fittings
  • Bronze self-lubricating bushings
  • Galvanized metal parts
  • Accommodates 1/2 in rope diameter
  • 2-1/4 in outside diameter sheave
  • 4 in block size
  • Double number of sheaves
  • Swivel hook with latch end fitting
  • 1400 lb working load limit