Learn More
MSC Campbell Eye Hook, 4,300 lb Capacity, Grade 100, Alloy Steel, 9/32 in Chain Dia
Catalog No. 5000527367
Encompass
The Campbell eye hook is designed for heavy-duty lifting applications, featuring a robust 4,300 lb capacity and a Grade 100 rating for superior strength. Constructed from durable alloy steel, it is suitable for demanding environments and ensures reliable performance in chain assemblies.
  • 4,300 lb capacity
  • Chain grade 100
  • Alloy steel construction
  • 0.37 in hook throat
  • 2-1/2 in reach
  • 0.56 in eye ID
  • 9/32 in chain diameter
  • 3.69 in overall length
Catalog No. 50-005-27367 Supplier MSC Supplier No. 67780924
The Campbell eye hook is designed for heavy-duty lifting applications, featuring a robust 4,300 lb capacity and a Grade 100 rating for superior strength. Constructed from durable alloy steel, it is suitable for demanding environments and ensures reliable performance in chain assemblies.
  • 4,300 lb capacity
  • Chain grade 100
  • Alloy steel construction
  • 0.37 in hook throat
  • 2-1/2 in reach
  • 0.56 in eye ID
  • 9/32 in chain diameter
  • 3.69 in overall length