Learn More
MSC Campbell Fixed Lifting Eye Bolt with Shoulder, 6,000 lb, 3/4-10, C-1030 Steel
Catalog No. 5000533927
Encompass
This fixed lifting eye bolt is designed for secure and reliable lifting applications. It features a shoulder for enhanced stability and is constructed from durable C-1030 forged steel. With a robust 6,000 lb lifting capacity, this bolt is suitable for a wide range of industrial and manufacturing tasks requiring dependable anchoring and lifting.
  • Fixed eye bolt type with shoulder
  • 6,000 lb lifting capacity
  • Eye inside diameter of 1-1/2 in
  • Eye outside diameter of 2-13/16 in
  • 3/4-10 thread size with 2 in fully threaded length
  • Overall length of 5-1/4 in and 2 in shank length
  • Constructed from C1030 forged steel
Catalog No. 50-005-33927 Supplier MSC Supplier No. 31638125
This fixed lifting eye bolt is designed for secure and reliable lifting applications. It features a shoulder for enhanced stability and is constructed from durable C-1030 forged steel. With a robust 6,000 lb lifting capacity, this bolt is suitable for a wide range of industrial and manufacturing tasks requiring dependable anchoring and lifting.
  • Fixed eye bolt type with shoulder
  • 6,000 lb lifting capacity
  • Eye inside diameter of 1-1/2 in
  • Eye outside diameter of 2-13/16 in
  • 3/4-10 thread size with 2 in fully threaded length
  • Overall length of 5-1/4 in and 2 in shank length
  • Constructed from C1030 forged steel