Learn More
MSC Campbell S-Hook, Trade Size #80, 160 Lb Capacity, Carbon Steel, Zinc Plated
Catalog No. 5000512516
Encompass
A durable S-hook designed for various hanging and connecting applications. It features a robust carbon steel construction with a zinc-plated finish for corrosion resistance.
  • 160 Lb load capacity
  • Constructed from carbon steel
  • Zinc-plated finish for enhanced durability
  • 1/4 in wire size
  • Overall length of 1-3/32 in
Catalog No. 50-005-12516 Supplier MSC Supplier No. 67788224
A durable S-hook designed for various hanging and connecting applications. It features a robust carbon steel construction with a zinc-plated finish for corrosion resistance.
  • 160 Lb load capacity
  • Constructed from carbon steel
  • Zinc-plated finish for enhanced durability
  • 1/4 in wire size
  • Overall length of 1-3/32 in