Learn More
MSC Campbell Single Jack Chain, No. 12, 100 ft, Zinc Finish
Catalog No. 5000536316
Encompass
This single jack chain is suitable for a variety of general-purpose applications. It is supplied in a 100 ft roll, providing ample length for numerous tasks, and features a zinc finish for enhanced durability and corrosion resistance. The chain has a consistent 0.125 in diameter and a reliable load capacity.
  • Single jack chain design
  • Durable zinc finish
  • 100 ft roll for extended use
  • 0.125 in chain diameter
  • 29 lb load capacity
Catalog No. 50-005-36316 Supplier MSC Supplier No. 18063214
This single jack chain is suitable for a variety of general-purpose applications. It is supplied in a 100 ft roll, providing ample length for numerous tasks, and features a zinc finish for enhanced durability and corrosion resistance. The chain has a consistent 0.125 in diameter and a reliable load capacity.
  • Single jack chain design
  • Durable zinc finish
  • 100 ft roll for extended use
  • 0.125 in chain diameter
  • 29 lb load capacity