Learn More
MSC Campbell Wire Rope Thimble Clip, 1/2 in Rope Diameter, Steel, Heavy-Galvanized
Catalog No. 5000537414
Encompass
This wire rope thimble clip is designed for a 1/2 in rope diameter, constructed from steel with a heavy-galvanized finish. It features a locking latch type and functions as a thimble fitting for secure rope applications.
  • Designed for 1/2 in rope diameter
  • Constructed from steel for durability
  • Heavy-galvanized finish provides corrosion resistance
  • Features a locking latch type
  • Functions as a thimble fitting for secure rope applications
  • Offers a loop inside width of 1.4375 in
  • Provides a clamp width of 2.6875 in
  • Backed by a hassle-free guarantee for customer satisfaction
Catalog No. 50-005-37414 Supplier MSC Supplier No. 67778944
This wire rope thimble clip is designed for a 1/2 in rope diameter, constructed from steel with a heavy-galvanized finish. It features a locking latch type and functions as a thimble fitting for secure rope applications.
  • Designed for 1/2 in rope diameter
  • Constructed from steel for durability
  • Heavy-galvanized finish provides corrosion resistance
  • Features a locking latch type
  • Functions as a thimble fitting for secure rope applications
  • Offers a loop inside width of 1.4375 in
  • Provides a clamp width of 2.6875 in
  • Backed by a hassle-free guarantee for customer satisfaction