Learn More
Cayman Chemical Campesterol 10mg
SDP
Catalog No. 502978493
Encompass_Preferred
A phytosterol with diverse biological activities; inhibits basic FGF-induced proliferation in, and vessel formation by, HUVECs at 1, 5, or 10 µM.; decreases the proliferation of MCF-7 and ZR-75-1 cells at 15 µM; reduces the levels of ERα in, and the diameter of, patient-derived breast cancer organoids at 10 µM; has been used as a marker of cholesterol intestinal absorption in humans
Catalog No. 50-297-8493 Supplier Cayman Chemical Supplier No. 4169510 MG
A phytosterol with diverse biological activities; inhibits basic FGF-induced proliferation in, and vessel formation by, HUVECs at 1, 5, or 10 µM.; decreases the proliferation of MCF-7 and ZR-75-1 cells at 15 µM; reduces the levels of ERα in, and the diameter of, patient-derived breast cancer organoids at 10 µM; has been used as a marker of cholesterol intestinal absorption in humans