Learn More
Kyfora Bio Campesterol 1 MG
Catalog No. 502559796
Encompass_Preferred
Campesterol is a naturally occurring phytosterol and cholesterol analog commonly used in lipid nanoparticle (LNP) formulations. Incorporating campesterol into LNPs has been shown to modify membrane structure, enhancing uptake efficiency in both in vitro and in vivo studies. This characteristic makes it a valuable component in the development of mRNA and nucleic acid delivery systems, where efficient cellular entry is crucial. For research use only.Key Applications: Lipid nanoparticles (LNPs), mRNA vaccine development
Catalog No. 50-255-9796 Supplier Kyfora Bio Supplier No. 145111
Campesterol is a naturally occurring phytosterol and cholesterol analog commonly used in lipid nanoparticle (LNP) formulations. Incorporating campesterol into LNPs has been shown to modify membrane structure, enhancing uptake efficiency in both in vitro and in vivo studies. This characteristic makes it a valuable component in the development of mRNA and nucleic acid delivery systems, where efficient cellular entry is crucial. For research use only.Key Applications: Lipid nanoparticles (LNPs), mRNA vaccine development