Learn More
Medchemexpress LLC HY-N1459 5mg , Campesterol CAS:474-62-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN14595MG
Encompass_Preferred
HY-N1459 5mg Campesterol CAS: 474-62-4 Campesterol is a plant sterol with cholesterol lowering and anticarcinogenic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1186