Learn More
TARGETMOL CHEMICALS INC CAMPESTEROL 5MG
SDP
Catalog No. 502576714
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Campesterol ((24R)-5-Ergosten-3(beta)-ol) is a plant sterol with cholesterol lowering and anticarcinogenic effects it and other plant sterols often decrease LDL cholesterol levels overall. Campesterol has anti-inflammatory effect it inhibits several pro-inflammatory and matrix degradation mediators typically involved in osteoarthritis- induced cartilage degradation also sometimes used to treat some specific prostate conditions. purity: 99%
Catalog No. 50-257-6714 Supplier TARGETMOL CHEMICALS INC Supplier No. T4S21575MG
Add to Cart
Add to Cart
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Campesterol ((24R)-5-Ergosten-3(beta)-ol) is a plant sterol with cholesterol lowering and anticarcinogenic effects it and other plant sterols often decrease LDL cholesterol levels overall. Campesterol has anti-inflammatory effect it inhibits several pro-inflammatory and matrix degradation mediators typically involved in osteoarthritis- induced cartilage degradation also sometimes used to treat some specific prostate conditions. purity: 99%