Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Camphene, MilliporeSigma™ Supelco™

Catalog No. 111020628 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0628 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0628 Supplier MilliporeSigma Supelco Supplier No. 442505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Camphene is an organic plastic type of crystal, which can be commonly used as an industrial intermediate compound for the production of commercial chemicals like camphor, isobornyl acetate and isoborneol. It can be obtained, via isomerization of alpha-pinene over acidic titanium dioxide catalyst.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 79-92-5
Density 0.85 g/mL (at 25°C)
Flash Point 29°C
Physical Form Neat
Linear Formula C10H16
MDL Number MFCD00066603
Melting Point 48°C to 52°C
Boiling Point 159°C to 160°C
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 136.23
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.