Learn More
Medchemexpress LLC (+)-camphor-10-sulfonic acid | 3144-16-9 | MFCD00064157 | ≥98.0% | 232.3 g·mol⁻¹ | C10H16O4S | 250g
SDP
Catalog No. 5000269286
Encompass_Preferred
( )-Camphor-10-sulfonic acid (( )-10-Camphorsulfonic acid) is an effective pancreatic lipase (PL) inhibitor ( )-Camphor-10-sulfonic acid is promising for research of obesity[1]
Catalog No. 50-002-69286 Supplier Medchemexpress LLC Supplier No. HYY0606250G
( )-Camphor-10-sulfonic acid (( )-10-Camphorsulfonic acid) is an effective pancreatic lipase (PL) inhibitor ( )-Camphor-10-sulfonic acid is promising for research of obesity[1]