Learn More
Medchemexpress LLC (±)-Camphor-10-sulfonic acid | 5872-08-2 | 99.8% | 232.30 | 100 G
SDP
Catalog No. 5000453829
Encompass_Preferred
(±)-Camphor-10-sulfonic acid is a racemate of Camphor-10-sulfonic acid. It functions as an organocatalyst, capable of facilitating a wide range of organic transformations. As a Brönsted acid catalyst, it plays a role in reducing the activation energy of chemical reactions by donating protons. It is known to catalyze various types of reactions, including Friedel-Crafts, Michael addition, and Mannich reactions.
  • Organocatalyst for diverse organic transformations.
  • Brönsted acid catalyst, which provides protons (H+) to reduce the activation energy of reactions.
  • Catalyzes Friedel-Crafts reaction, Michael addition reaction, and Mannich reaction.
Catalog No. 50-004-53829 Supplier Medchemexpress LLC Supplier No. HYW012218100G
(±)-Camphor-10-sulfonic acid is a racemate of Camphor-10-sulfonic acid. It functions as an organocatalyst, capable of facilitating a wide range of organic transformations. As a Brönsted acid catalyst, it plays a role in reducing the activation energy of chemical reactions by donating protons. It is known to catalyze various types of reactions, including Friedel-Crafts, Michael addition, and Mannich reactions.
  • Organocatalyst for diverse organic transformations.
  • Brönsted acid catalyst, which provides protons (H+) to reduce the activation energy of reactions.
  • Catalyzes Friedel-Crafts reaction, Michael addition reaction, and Mannich reaction.