Learn More
Medchemexpress LLC Camphor | 76-22-2 | 152.24 | 25 G
SDP
Catalog No. 5000389612
Encompass_Preferred
Camphor ((±)-Camphor) is a chemical compound known for its topical anti-infective and anti-pruritic properties. Internally, it acts as a stimulant and carminative. It is important to note that camphor is poisonous when ingested. It also exhibits various biological activities.
  • Exhibits antiviral activities
  • Exhibits antitussive activities
  • Exhibits anticancer activities
  • Identified as a TRPV3 agonist
Catalog No. 50-003-89612 Supplier Medchemexpress LLC Supplier No. 25GHYN0808
Camphor ((±)-Camphor) is a chemical compound known for its topical anti-infective and anti-pruritic properties. Internally, it acts as a stimulant and carminative. It is important to note that camphor is poisonous when ingested. It also exhibits various biological activities.
  • Exhibits antiviral activities
  • Exhibits antitussive activities
  • Exhibits anticancer activities
  • Identified as a TRPV3 agonist