Learn More
Medchemexpress LLC Camphor (Standard) | 76-22-2 | MFCD00074738 | 152.23 | 100 MG
SDP
Catalog No. 5000416685
Encompass_Preferred
Camphor (Standard) is an analytical standard intended for research and analytical applications. It acts as a topical anti-infective and anti-pruritic, and internally as a stimulant and carminative. It exhibits antiviral, antitussive, and anticancer activities, and is also a TRPV3 agonist.
  • Analytical standard for research and analytical applications.
  • Topical anti-infective and anti-pruritic.
  • Internally acts as a stimulant and carminative.
  • Exhibits antiviral, antitussive, and anticancer activities.
  • TRPV3 agonist.
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-004-16685 Supplier Medchemexpress LLC Supplier No. HYN0808R100MG
Camphor (Standard) is an analytical standard intended for research and analytical applications. It acts as a topical anti-infective and anti-pruritic, and internally as a stimulant and carminative. It exhibits antiviral, antitussive, and anticancer activities, and is also a TRPV3 agonist.
  • Analytical standard for research and analytical applications.
  • Topical anti-infective and anti-pruritic.
  • Internally acts as a stimulant and carminative.
  • Exhibits antiviral, antitussive, and anticancer activities.
  • TRPV3 agonist.
  • Used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.