Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Camphor sulfonic acid methyl ester | 62319-13-5 | MFCD29045182 | 246.32 | C11H18O4S | 25 MG
SDP

Catalog No. 5000284873
Encompass_Preferred

Camphor sulfonic acid methyl ester is supplied as a research reagent and impurity standard for use in analytical and synthetic workflows. It is the methyl ester of camphorsulfonic acid (Methyl (1S)-(+)-10-camphorsulfonate) and is provided with storage instructions in the certificate of analysis.

  • Contains methyl (1S)-(+)-10-camphorsulfonate (camphor sulfonic acid methyl ester).
  • CAS number 62319-13-5.
  • Chemical formula C11H18O4S; molecular weight 246.32.
  • Intended for use as a camphor impurity or drug intermediate in research.
  • Available in small pack sizes suitable for analytical and synthetic work (for example, 25 MG).
  • Store under conditions specified in the certificate of analysis.

Catalog No. 50-002-84873 Supplier Medchemexpress LLC Supplier No. HYW58740525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Camphor sulfonic acid methyl ester is supplied as a research reagent and impurity standard for use in analytical and synthetic workflows. It is the methyl ester of camphorsulfonic acid (Methyl (1S)-(+)-10-camphorsulfonate) and is provided with storage instructions in the certificate of analysis.

  • Contains methyl (1S)-(+)-10-camphorsulfonate (camphor sulfonic acid methyl ester).
  • CAS number 62319-13-5.
  • Chemical formula C11H18O4S; molecular weight 246.32.
  • Intended for use as a camphor impurity or drug intermediate in research.
  • Available in small pack sizes suitable for analytical and synthetic work (for example, 25 MG).
  • Store under conditions specified in the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.