Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Camphor sulfonic acid methyl ester | 62319-13-5 | MFCD29045182 | 246.32 | C11H18O4S | 500 MG
SDP

Catalog No. 5000284874
Encompass_Preferred

Camphor sulfonic acid methyl ester is a camphor-derived sulfonate ester used as a drug intermediate and an analytical impurity reference. It is described as Methyl (1S)-(+)-10-camphorsulfonate (CAS 62319-13-5) with formula C11H18O4S and molecular weight 246.32; storage and purity details are provided in the product Certificate of Analysis.

  • Used as a drug intermediate and synthetic building block.
  • Serves as an impurity or analytical reference for camphor-related analyses.
  • Molecular formula C11H18O4S and molecular weight 246.32 support formulation and calculation needs.
  • CAS-registered substance (CAS 62319-13-5) for unambiguous identification.
  • Storage and assay information available in the certificate of analysis for each batch.

Catalog No. 50-002-84874 Supplier Medchemexpress LLC Supplier No. HYW587405500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Camphor sulfonic acid methyl ester is a camphor-derived sulfonate ester used as a drug intermediate and an analytical impurity reference. It is described as Methyl (1S)-(+)-10-camphorsulfonate (CAS 62319-13-5) with formula C11H18O4S and molecular weight 246.32; storage and purity details are provided in the product Certificate of Analysis.

  • Used as a drug intermediate and synthetic building block.
  • Serves as an impurity or analytical reference for camphor-related analyses.
  • Molecular formula C11H18O4S and molecular weight 246.32 support formulation and calculation needs.
  • CAS-registered substance (CAS 62319-13-5) for unambiguous identification.
  • Storage and assay information available in the certificate of analysis for each batch.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.