Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Camphor sulfonic acid methyl ester | 62319-13-5 | MFCD29045182 | >95.0% | 246.32 | C11H18O4S | 100 MG
SDP

Catalog No. 5000280471
Encompass_Preferred

Camphor sulfonic acid methyl ester (Methyl (1S)-(+)-10-camphorsulfonate) is a methyl ester impurity of camphor used as a reference standard and research reagent for analytical and synthetic chemistry. It is identified by CAS 62319-13-5 and characterized by its molecular formula and mass.

  • Used as a reference impurity in analytical characterization.
  • CAS number 62319-13-5 for clear identification.
  • Molecular formula C11H18O4S, molecular weight 246.32 g/mol.
  • Available in 25 mg-10 g pack sizes suitable for research.
  • Purity typically reported >95%; confirm certificate of analysis before use.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-80471 Supplier Medchemexpress LLC Supplier No. HYW587405100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Camphor sulfonic acid methyl ester (Methyl (1S)-(+)-10-camphorsulfonate) is a methyl ester impurity of camphor used as a reference standard and research reagent for analytical and synthetic chemistry. It is identified by CAS 62319-13-5 and characterized by its molecular formula and mass.

  • Used as a reference impurity in analytical characterization.
  • CAS number 62319-13-5 for clear identification.
  • Molecular formula C11H18O4S, molecular weight 246.32 g/mol.
  • Available in 25 mg-10 g pack sizes suitable for research.
  • Purity typically reported >95%; confirm certificate of analysis before use.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.