Learn More
AdipoGen Camphoric acid
SDP
Catalog No. 502561455
Encompass_Preferred
Chemical. CAS 560-09-8. Formula C10H16O4. MW 200.23. Synthetic Chirality inducing agent. Useful tool as resolving agent to separate the isomers and purify racemic mixtures e.g. DL-lysine by fractionally crystallizing the product to obtain the pure isomer e.g. D-lysine. Can also be used as building block.
Catalog No. 50-256-1455 Supplier AdipoGen Supplier No. CDXC0144G010
Chemical. CAS 560-09-8. Formula C10H16O4. MW 200.23. Synthetic Chirality inducing agent. Useful tool as resolving agent to separate the isomers and purify racemic mixtures e.g. DL-lysine by fractionally crystallizing the product to obtain the pure isomer e.g. D-lysine. Can also be used as building block.