Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Camphorquinone | 10373-78-1 | MFCD00064160 | ≥97.0% | 166.22 | C10H14O2 | 25 G
SDP

Catalog No. 5000209017
Encompass_Preferred

Camphorquinone is a visible-light photoinitiator used to generate free radicals for polymerization in dental resins and coatings. It is supplied as a light yellow to yellow solid with formula C10H14O2 and a typical purity of ≥97.0%.

  • photoinitiator for visible-light polymerization
  • light yellow to yellow solid appearance
  • molecular weight 166.22 g/mol
  • purity ≥97.0%
  • store powder at -20°C for long-term stability

Catalog No. 50-002-09017 Supplier Medchemexpress LLC Supplier No. 25GHYW086630
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Camphorquinone is a visible-light photoinitiator used to generate free radicals for polymerization in dental resins and coatings. It is supplied as a light yellow to yellow solid with formula C10H14O2 and a typical purity of ≥97.0%.

  • photoinitiator for visible-light polymerization
  • light yellow to yellow solid appearance
  • molecular weight 166.22 g/mol
  • purity ≥97.0%
  • store powder at -20°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.