Learn More
AdipoGen Camphorylsulfonyl oxaziridine
SDP
Supplier:  AdipoGen CDXD0021G005
Encompass_Preferred
Chemical. CAS 139628-16-3. Formula C10H13Cl2NO3S. MW 298.19. Synthetic. Chiral oxidizing agent. Used for asymmetric synthesis as chiral catalyst. Hydroxylation reagent.
Catalog No. 50-256-1314