Learn More
Bioworld Camptothecin, 1 g
Supplier:  Bioworld 403001493
Encompass_Preferred
A reversible Mitochondrial Topo I inhibitor shown to exhibit antitumour activityAppearance: SolidBoiling Point : 757.01 °C at 760 mmHgDensity : 1.51 g/cm3Melting Point: 260 °C (dec.)(lit.)RTECS Number: UQ0492000
Catalog No. 50-255-695