Learn More
Medchemexpress LLC Camptothecin-20S-O-p 25mg | 194414-69-2 | 25 MG
SDP
Catalog No. 5000229254
Encompass_Preferred
Camptothecin-20(S)-O-propionate (CZ48) is the C20-propionate ester of camptothecin that functions as a topoisomerase I inhibitor and has reported antitumor activity in animal models. It is provided as a solid and as prepared solutions for research use, with recommended storage to maintain stability.
  • Acts as a topoisomerase I inhibitor
  • Demonstrated antitumor activity in breast and prostate carcinoma models
  • Available as solid and 10 mM DMSO solution formulations
  • High purity suitable for research (reported ~99.5%)
  • Stable under recommended storage: powder -20°C for long term, solvent -80°C
Catalog No. 50-002-29254 Supplier Medchemexpress LLC Supplier No. HY11466825MG
Camptothecin-20(S)-O-propionate (CZ48) is the C20-propionate ester of camptothecin that functions as a topoisomerase I inhibitor and has reported antitumor activity in animal models. It is provided as a solid and as prepared solutions for research use, with recommended storage to maintain stability.
  • Acts as a topoisomerase I inhibitor
  • Demonstrated antitumor activity in breast and prostate carcinoma models
  • Available as solid and 10 mM DMSO solution formulations
  • High purity suitable for research (reported ~99.5%)
  • Stable under recommended storage: powder -20°C for long term, solvent -80°C